Please login to your online account to display your discounted pricing

Science FUNdamentals DVDs


Creative videos correlated to national standards.

Manufacturer: fisher science education  KF520

 View more versions of this product

Catalog No. S66450

  • / Each
 

Description & Specifications

Specifications

Includes Marc Record, Teacher's Guide
Grade 4 to 8
Format VHS
Name Magic Light Show-What is Light? VHS
Copyright Year 2000