missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

TMW Media Science FUNdamentals DVDs

Creative videos correlated to national standards.

Manufacturer:  TMW Media KF524

 View more versions of this product

Catalog No. S66449



Description

Description

Each video includes a teacher's guide with standards correlations, vocabulary terms, sample support activities, and study questions. Run time: 15 min.

Gravity Is Attractive: What Is Gravity?
Learn what gravity is and how it affects our physical world. Students will see experiments and activities that explain gravity as a property of matter and basic force.
The Magic Light Show: What Is Light?
Explore the “magic” of light and learn amazing facts about how we see things. Features the basic principles of light, light transmission, and reflection and refraction in different materials.
The Earth Is a Giant Magnet: Understanding Magnetism
Learn about the invisible force of magnetism. Demonstrates that the Earth is a giant magnet and illustrates the basic principles of magnetism.
Newton's Wagon: Understanding Motion
Learn about Newton's three laws of motion and how they apply to everything we do.
Let's Form a Band: Understanding Sound
Discover the principles of sound and help make your own band. Teaches the basic terms such as amplitude, frequency pitch, and timbre.
Getting Charged: Understanding Static Electricity
Learn the principles of electrostatics and explore how electrons move.

Specifications

Specifications

2000
Gravity is Attractive-What is Gravity? VHS
8
Education
Marc Record, Teacher's Guide
VHS
4
Documents