Science First™ Spectrum Tubes
Tubes produce bright, well-defined, unique spectra when the gas inside is excited before a spectroscope.
Supplier: Science First™ 6140745
Requires : Spectrum tube power supply (S05966)
Description
Fit easily into Spectrum Tube Power Supply (S05966).
Glass construction9.5 in. long and narrowed to capillary width for about 3.9 in. (10cm) in the center
Sealed metal wires hold the electrodes and are welded to metal caps which have a loop for convenient hookup to wire connections
Specifications
Spectrum Tube, Helium | |
9.5 in. | |
3.9 in. |
9.5 x 3.9 in. (24.13 x 9.91 cm) | |
24.13 cm | |
9.91 cm |