Please login to your online account to display your discounted pricing

Science for Every Kid Series

Hands-on projects every student will love

Manufacturer: fisher science education  0471620858

 View all options for this product

Catalog No. S51357-1


Description & Specifications


Author Janice VanCleave
Category Chemistry
Format Book
Grade 2 to 7
Product Type Chemistry For Every Kid: 101 Easy Experiments that Really Work

Janice VanCleave books contain 100 easy and low-cost experiments that really work. Each experiment includes a statement of purpose, materials list, instructions and discussion. Save on the complete set.