Please login to your online account to display your discounted pricing

Science Challenge Series

Motivates students through high-interest game format.

Manufacturer: fisher science education  #04342X

 View all options for this product

Catalog No. S66859


Description & Specifications


Category Chemistry
For Use With (Application) Students with low reading levels
Format Book
Product Type Student Chemistry Book

  • Challenges all students with five levels of difficulty
  • Correlates to major science textbooks and National Science Education Standards