Please login to your online account to display your discounted pricing

SCH 79797 dihydrochloride, Tocris

Potent, selective non-peptide PAR1 antagonist

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material SCH 79797 dihydrochloride
Assay Percent Range >99%
Molecular Formula C23H25N5·2HCl
Formula Weight 444.41
Synonym N3-Cyclopropyl-7-[[4-(1-methylethyl)phenyl]methyl]-7H-pyrrolo[3, 2-f]quinazoline-1, 3-diamine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

159250

 
r & d systems
159250
50mg Each for $1,205.80

159210

 
r & d systems
159210
10mg Each for $290.95
Description & Specifications

Specifications

Formula Weight 444.41
Quantity 50mg
Chemical Name or Material SCH 79797 dihydrochloride
Assay Percent Range >99%
Molecular Formula C23H25N5·2HCl
CAS 1216720-69-2
Synonym N3-Cyclopropyl-7-[[4-(1-methylethyl)phenyl]methyl]-7H-pyrrolo[3, 2-f]quinazoline-1, 3-diamine dihydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature