Please login to your online account to display your discounted pricing

SCH 529074, Tocris™

Restores mutant p53 activity

$251.85 - $1,045.45

Chemical Identifiers

Chemical Name or Material SCH 529074
Assay Percent Range >98%
Molecular Formula C31H36Cl2N6
Formula Weight 563.56
Synonym N3-[2-[[4-[Bis(4-chlorophenyl)methyl]-1-piperazinyl]methyl]-4-quinazolinyl]-N1, N1-dimethyl-1, 3-propanediamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

424010

 
r & d systems
424010
10mg Each for $251.85

424050

 
r & d systems
424050
50mg Each for $1,045.45
Description & Specifications

Specifications

Formula Weight 563.56
Quantity 10mg
Chemical Name or Material SCH 529074
Assay Percent Range >98%
Molecular Formula C31H36Cl2N6
CAS 922150-11-6
Synonym N3-[2-[[4-[Bis(4-chlorophenyl)methyl]-1-piperazinyl]methyl]-4-quinazolinyl]-N1, N1-dimethyl-1, 3-propanediamine
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C