Please login to your online account to display your discounted pricing

SCH 50911, Tocris


Selective, competitive, orally active GABAB antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS098410

 View more versions of this product

Catalog No. 09-841-0

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 173.21
CAS 733717-87-8
Assay Percent Range >98%
Synonym (2S)-(+)-5, 5-Dimethyl-2-morpholineacetic acid
Recommended Storage Desiccate at -20°C
Molecular Formula C8H15NO3
Chemical Name or Material SCH 50911

Specifications

Chemical Name or Material SCH 50911
Assay Percent Range >98%
Molecular Formula C8H15NO3
Formula Weight 173.21
Synonym (2S)-(+)-5, 5-Dimethyl-2-morpholineacetic acid
Accessories

Desiccate at -20°C