Please login to your online account to display your discounted pricing

SCH 23390 hydrochloride, Tocris


Standard selective D1-like antagonist; Also 5-HT2C agonist

Quantity 10mg

Manufacturer: r & d systems  092510

 View more versions of this product

Catalog No. 092510

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 324.24
Chemical Name or Material SCH 23390 hydrochloride
Molecular Formula C17H18ClNO·HCl
Synonym (R)-(+)-7-Chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride
Recommended Storage Desiccate at +4°C
Assay Percent Range >98%
CAS 125941-87-9

Specifications

Chemical Name or Material SCH 23390 hydrochloride
Assay Percent Range >98%
Molecular Formula C17H18ClNO·HCl
Formula Weight 324.24
Synonym (R)-(+)-7-Chloro-8-hydroxy-3-methyl-1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine hydrochloride
Accessories

Desiccate at +4°C