Please login to your online account to display your discounted pricing

SCH 221510, Tocris

Potent and selective NOP agonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material SCH 221510
Assay Percent Range >99%
Molecular Formula C28H31NO
Formula Weight 397.55
Synonym 3-Endo-8-[bis(2-methylphenyl)methyl]-3-phenyl-8-azabicyclo[3.2.1]octan-3-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

324050

 
r & d systems
324050
50mg Each for $1,050.42

324010

 
r & d systems
324010
10mg Each for $252.39
Description & Specifications

Specifications

Chemical Name or Material SCH 221510
CAS 322473-89-2
Recommended Storage Store at Room Temperature
Assay Percent Range >99%
Molecular Formula C28H31NO
Formula Weight 397.55
Synonym 3-Endo-8-[bis(2-methylphenyl)methyl]-3-phenyl-8-azabicyclo[3.2.1]octan-3-ol
Quantity 50mg
Safety and Handling

Store at Room Temperature