Please login to your online account to display your discounted pricing

SCH 202676 hydrobromide, Tocris

Inhibitor of ligand binding to G-protein-coupled receptors

$214.12 - $895.76

Chemical Identifiers

Chemical Name or Material SCH 202676 hydrobromide
Assay Percent Range >99%
Molecular Formula C15H13N3S·HBr
Formula Weight 348.26
Synonym N-(2,3-Diphenyl-1,2,4-thiadiazol-5(2H)-ylidene)methanamine hydrobromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

140050

 
r & d systems
RDS140050
50mg Each for $895.76

140010

 
r & d systems
RDS140010
10mg Each for $214.12
Description & Specifications

Specifications

Formula Weight 348.26
Quantity 50mg
Chemical Name or Material SCH 202676 hydrobromide
Assay Percent Range >99%
CAS 265980-25-4
Synonym N-(2,3-Diphenyl-1,2,4-thiadiazol-5(2H)-ylidene)methanamine hydrobromide
Recommended Storage Desiccate at +4°C
Molecular Formula C15H13N3S·HBr
Safety and Handling

Desiccate at +4°C