Please login to your online account to display your discounted pricing

Scandium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 125mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  PLSC23Y

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. PLSC2-3Y

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 125mL
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Health Hazard 1 Causes skin irritation, serious eye irritation
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
DOT Information Hazardous
Solvent or Matrix 5% HNO3
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
CAS 12060-08-1
Chemical Name or Material Scandium Standard
CAS 7697-37-2
CAS 7732-18-5

Specifications

Chemical Name or Material Scandium Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Sc10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes skin irritation, serious eye irritation

Ambient