Please login to your online account to display your discounted pricing

Alfa Aesar™ Scandium, plasma standard solution, Specpure™, Sc 10,000μg/mL

Chemical Identifiers

Chemical Name or Material Scandium plasma standard solution
Name Note Sc 10000μg/mL
Purity Grade Specpure™
MDL Number MFCD00016323
UN Number UN3264
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35755AD

  SDS
alfa aesar™
35755AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35755AE

  SDS
alfa aesar™
35755AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35755AP

  SDS
alfa aesar™
35755AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Scandium plasma standard solution
Quantity 50mL
TSCA TSCA
Name Note Sc 10000μg/mL
Purity Grade Specpure™
Physical Form Liquid
Concentration Sc2O3 in 5% HNO3
MDL Number MFCD00016323
UN Number UN3264
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.