missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scandium ingot, 99.9% (metals basis excluding Ta), Ta nominal 8-12%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04022903

 View more versions of this product

Catalog No. AA4022903



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Specifications

Gray
7440-20-2
Sc
1 g
SIXSYDAISGFNSX-UHFFFAOYSA-N
scandium
23952
99.9%
Ingot
Scandium ingot
(metals basis excluding Ta), Ta nominal 8 to 12%
MFCD00016323
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
[Sc]
44.96
CHEBI:33330
Odorless

Chemical Identifiers

7440-20-2
44.96
SIXSYDAISGFNSX-UHFFFAOYSA-N
23952
scandium
Sc
MFCD00016323
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
CHEBI:33330
[Sc]
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : Hazard Class: 4.1; Packaging Group: III

EINECSNumber : 231-129-2

TSCA : Yes

storageNote1 :

  • Packaged under argon

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only