missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scandium(III) Trifluoromethanesulfonate, TCI America™

Manufacturer:  TCI America T16631G

 View more versions of this product

Catalog No. T16631G



Specifications

Specifications

Scandium(III) Trifluoromethanesulfonate
1g
C3F9O9S3Sc
scandium iii trifluoromethanesulfonate, scandium trifluoromethanesulfonate, scandium iii triflate, scandium triflate, sc otf 3, scandium 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid scandium iii salt, scandium iii trifluoromethanesulphonate, scandium tris trifluoromethanesulfonate, scandiumtriflate
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]
492.146
492.15
≥98.0% (T)
White
144026-79-9
MFCD00192433
HZXJVDYQRYYYOR-UHFFFAOYSA-K
scandium(3+);trifluoromethanesulfonate
2734571
Crystal-Powder at 20°C

Chemical Identifiers

144026-79-9
492.146
HZXJVDYQRYYYOR-UHFFFAOYSA-K
2734571
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Sc+3]
C3F9O9S3Sc
MFCD00192433
scandium iii trifluoromethanesulfonate, scandium trifluoromethanesulfonate, scandium iii triflate, scandium triflate, sc otf 3, scandium 3+ ; trifluoromethanesulfonate, trifluoromethanesulfonic acid scandium iii salt, scandium iii trifluoromethanesulphonate, scandium tris trifluoromethanesulfonate, scandiumtriflate
scandium(3+);trifluoromethanesulfonate
Safety and Handling

Safety and Handling

TSCA : No

Documents