Please login to your online account to display your discounted pricing

Scandium(III) oxide, 99.99% (REO), Alfa Aesar™, Reacton™

Chemical Identifiers

Chemical Name or Material Scandium(III) oxide
CAS 12060-08-1
Physical Form Powder
Assay Percent Range 99.99%
Molecular Formula Sc2O3
Formula Weight 137.91
MDL Number MFCD00011223
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1121706

  SDS
alfa aesar
1121706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1121714

  SDS
alfa aesar
1121714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1121703

  SDS
alfa aesar
1121703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 137.91
Quantity 5g
EINECS Number 235-042-0
Physical Form Powder
Molecular Formula Sc2O3
Odor Odorless
Assay Percent Range 99.99%
CAS 12060-08-1
MDL Number MFCD00011223
Chemical Name or Material Scandium(III) oxide