missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scandium(III) oxide, 99.99% (REO), Alfa Aesar™, Reacton™

Manufacturer:  Alfa Aesar 1121714

 View more versions of this product

Catalog No. AA1121714



Specifications

Specifications

Scandium(III) oxide
99.99%
MFCD00011223
Powder
25g
12060-08-1
Sc2O3
137.91
Odorless

Chemical Identifiers

12060-08-1
Sc2O3
MFCD00011223
Safety and Handling

Safety and Handling

EINECSNumber : 235-042-0

Documents