Learn More
Scandium(III) nitrate hydrate, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 06710406
Description
Scandium(III) nitrate hydrate is used as an oxidizing agent in organic synthesis. It is used in the optical coatings, electronic ceramics and laser industries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Scandium (III) nitrate hydrate | |
White | |
5 g | |
N3O9Sc | |
UN1477 | |
scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium salt | |
DFCYEXJMCFQPPA-UHFFFAOYSA-N | |
scandium(3+);trinitrate | |
166818 | |
99.9% | |
Hygroscopic |
107552-14-7 | |
Crystalline | |
(REO) | |
MFCD00011220 | |
14,8392 | |
Soluble in water and alcohol. | |
[Sc+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
230.97 | |
230.97 (Anhydrous) | |
Reacton™ |
Chemical Identifiers
107552-14-7 | |
MFCD00011220 | |
scandium nitrate, scandium iii nitrate, pa4cjgprtn, unii-pa4cjgprtn, scandium iii nitrate pentahydrate-sc reo, nitric acid, scandium 3+ salt 3:1, scandium 3+ trinitrate, acmc-1bol9, nitric acid, scandium salt | |
scandium(3+);trinitrate |
N3O9Sc | |
DFCYEXJMCFQPPA-UHFFFAOYSA-N | |
166818 |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H272-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S.
EINECSNumber : 236-701-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only