Learn More
Alfa Aesar™ Scandium(III) iodide, ultra dry, 99.999% (REO)
Manufacturer: Alfa Aesar™ 1368703
Description
- Ampouled under argon
Specifications
Scandium(III) iodide | |
14474-33-0 | |
I3Sc | |
scandium iodide, scandium iii iodide, unii-qvj18sk04t, qvj18sk04t, scandium 3+ triiodide, scandium iii triiodide, acmc-1aq4e | |
[Sc+3].[I-].[I-].[I-] | |
425.669 | |
425.67 | |
99.999% | |
Hygroscopic | |
1g |
Ultra dry | |
(REO) | |
MFCD00054037 | |
HUIHCQPFSRNMNM-UHFFFAOYSA-K | |
scandium(3+);triiodide | |
84464 | |
Powder | |
Ampule | |
945°C |
Chemical Identifiers
14474-33-0 | |
425.669 | |
HUIHCQPFSRNMNM-UHFFFAOYSA-K | |
84464 | |
[Sc+3].[I-].[I-].[I-] |
I3Sc | |
MFCD00054037 | |
scandium iodide, scandium iii iodide, unii-qvj18sk04t, qvj18sk04t, scandium 3+ triiodide, scandium iii triiodide, acmc-1aq4e | |
scandium(3+);triiodide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
TSCA : TSCA
storageNote1 :
- Ampouled under argon