Please login to your online account to display your discounted pricing

Alfa Aesar™ Scandium(III) chloride hexahydrate, REacton™, 99.99% (REO)

Chemical Identifiers

Chemical Name or Material Scandium(III) chloride hexahydrate
CAS 20662-14-0
Purity Grade Reacton™
Formula Weight 259.41 (151.32 Anhydrous)
MDL Number MFCD00149852
Merck Index 14,8392
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1121903

  SDS
alfa aesar™
1121903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1121906

  SDS
alfa aesar™
1121906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 259.41 (151.32 Anhydrous)
Assay 99.99%
Chemical Name or Material Scandium(III) chloride hexahydrate
Sensitivity Hygroscopic
MDL Number MFCD00149852
CAS 20662-14-0
Physical Form Crystalline Aggregates
TSCA TSCA
Quantity 1g
Merck Index 14,8392
Purity Grade Reacton™
Assay Percent Notes (REO)