Please login to your online account to display your discounted pricing

Alfa Aesar™ Scandium(III) chloride hexahydrate, REacton™, 99.9% (REO)

Chemical Identifiers

Chemical Name or Material Scandium(III) chloride hexahydrate
CAS 20662-14-0
Purity Grade Reacton™
Formula Weight 259.41 (151.32 Anhydrous)
MDL Number MFCD00149852
Merck Index 14,8392
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1121803

 
alfa aesar™
1121803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1121806

 
alfa aesar™
1121806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 259.41 (151.32 Anhydrous)
Chemical Name or Material Scandium(III) chloride hexahydrate
TSCA TSCA
Purity Grade Reacton™
Physical Form Crystalline Aggregates
Assay 99.9%
Sensitivity Hygroscopic
Assay Percent Notes (REO)
MDL Number MFCD00149852
CAS 20662-14-0
Merck Index 14,8392
Quantity 1g