missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Scandium(III) bis(trifluoromethylsulfonyl)imide

Manufacturer:  Alfa Aesar™ 46948MD

 View more versions of this product

Catalog No. AA46948MD



Specifications

Specifications

Scandium (III) bis (trifluoromethylsulfonyl)imide
White to Yellow
250mg
C6F18N3O12S6Sc
MFCD03427000
scandium iii trifluoromethanesulfonimide
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sc+3]
885.362
885.37
Moisture Sensitive
199°C to 204°C
176726-07-1
[(CF3SO2)2N]3Sc
UN1759
FUXLYEZEIZAKTL-UHFFFAOYSA-N
bis(trifluoromethylsulfonyl)azanide;scandium(3+)
131875098
Solid

Chemical Identifiers

176726-07-1
885.362
FUXLYEZEIZAKTL-UHFFFAOYSA-N
131875098
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Sc+3]
C6F18N3O12S6Sc
MFCD03427000
scandium iii trifluoromethanesulfonimide
bis(trifluoromethylsulfonyl)azanide;scandium(3+)
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

SDS Documents