missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scandium(III) Acetate Hydrate 98.0+%, TCI America™

Manufacturer:  TCI America S05345G

 View more versions of this product

Catalog No. S05345G



Specifications

Specifications

Scandium(III) Acetate Hydrate
5g
C6H9O6Sc
Acetic Acid Scandium(III) Salt
Crystal-Powder at 20°C
White
207671-51-0
MFCD00150122
222.09
≥98.0% (T)

Chemical Identifiers

207671-51-0
C6H9O6Sc
MFCD00150122
Acetic Acid Scandium(III) Salt
Safety and Handling

Safety and Handling

TSCA : No

Documents