missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scandium(III) 2,4-pentanedionate hydrate, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4444803

 View more versions of this product

Catalog No. AA4444803



Specifications

Specifications

Scandium(III) 2, 4-pentanedionate hydrate
C15H23O7Sc
tris 4-hydroxypent-3-en-2-one hydrate scandium
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Sc+3]
360.298
342.26 (Anhydrous)
Odorless
174°C to 186°C
699012-88-9
MFCD00064715
LDRQBGBMJVMLGE-KJVLTGTBSA-K
(Z)-4-oxopent-2-en-2-olate;scandium(3+);hydrate
119030428
Powder
Colorless
1g

Chemical Identifiers

699012-88-9
360.298
LDRQBGBMJVMLGE-KJVLTGTBSA-K
119030428
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Sc+3]
C15H23O7Sc
MFCD00064715
tris 4-hydroxypent-3-en-2-one hydrate scandium
(Z)-4-oxopent-2-en-2-olate;scandium(3+);hydrate
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 238-194-6

TSCA : No

Recommended Storage : Ambient temperatures

Documents