missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Scandium ICP Standard, 1mL = 1mg Sc (1,000ppm Sc), Ricca Chemical

Manufacturer:  Ricca Chemical Company RIPSC1KN500

 View more versions of this product

Catalog No. PSC1KN-500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Scandium ICP Standard
93.0,5.34,0.14
Liquid
1,000ppm Sc
Colorless
12060-08-1,7697-37-2,7732-18-5
96.0,5.56,0.15
Poly natural
1 mL = 1 mg Sc
500mL

Chemical Identifiers

12060-08-1
Documents