missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Scandium foil, 0.25mm (0.01 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 12424FI

 View more versions of this product

Catalog No. AA12424FI



Specifications

Specifications

Scandium foil
50 x 50mm
7440-20-2
Sc
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
[Sc]
Odorless
Mylar bag
CHEBI:33330
99.9%
≈0.5g/25x25mm
0.25mm (0.01 in.) thick
(REO)
MFCD00016323
SIXSYDAISGFNSX-UHFFFAOYSA-N
scandium
44.956
23952
Foil

Chemical Identifiers

7440-20-2
44.956
SIXSYDAISGFNSX-UHFFFAOYSA-N
23952
scandium
Sc
MFCD00016323
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
CHEBI:33330
[Sc]
Safety and Handling

Safety and Handling

TSCA : TSCA

Documents