missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Scandium foil, 0.025mm (0.001 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 12426FI

 View more versions of this product

Catalog No. AA12426FI



Description

Description

  • Packaged under argon
Specifications

Specifications

Scandium foil
50 x 50mm
7440-20-2
Sc
UN3178
SIXSYDAISGFNSX-UHFFFAOYSA-N
scandium
44.956
CHEBI:33330
99.9%
≈0.05g/25x25mm
0.025mm (0.001 in.) thick
(REO)
MFCD00016323
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
[Sc]
Odorless
23952
Foil
Gray

Chemical Identifiers

7440-20-2
44.956
SIXSYDAISGFNSX-UHFFFAOYSA-N
23952
scandium
Sc
MFCD00016323
unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
CHEBI:33330
[Sc]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Documents