missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Scandium AA Standard, 1000 ppm Sc in 3% HNO3, Ricca Chemical
Supplier: Ricca Chemical Company ASC1KN500
Specifications
Scandium AA Standard | |
Natural Poly Bottle | |
92.0,5.54,0.15 | |
Colorless | |
Odorless |
12060-08-1,7697-37-2,7732-18-5 | |
500 mL | |
96.0,5.77,0.15 | |
1mL = 1mg Sc (1, 000ppm Sc)Sc2O3 in 3% HNO3 |
Chemical Identifiers
12060-08-1 |
Safety and Handling
ShelfLife : 24 months