Please login to your online account to display your discounted pricing

SC-9, Tocris

Protein kinase C activator

$177.37 - $746.48

Chemical Identifiers

Chemical Name or Material SC-9
Assay Percent Range >99%
Molecular Formula C22H24ClNO2S
Formula Weight 401.95
Synonym 5-Chloro-N-(6-phenylhexyl)-1-naphthalenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

043310

 
r & d systems
RDS043310
10mg Each for $177.37

043350

 
r & d systems
RDS043350
50mg Each for $746.48
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 401.95
Chemical Name or Material SC-9
CAS 102649-78-5
Molecular Formula C22H24ClNO2S
Assay Percent Range >99%
Synonym 5-Chloro-N-(6-phenylhexyl)-1-naphthalenesulfonamide
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature