Please login to your online account to display your discounted pricing

SC-9, Tocris


Protein kinase C activator

Quantity 50mg

Manufacturer: r & d systems  RDS043350

 View more versions of this product

Catalog No. 043350

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 401.95
Chemical Name or Material SC-9
CAS 102649-78-5
Assay Percent Range >99%
Molecular Formula C22H24ClNO2S
Synonym 5-Chloro-N-(6-phenylhexyl)-1-naphthalenesulfonamide
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material SC-9
Assay Percent Range >99%
Molecular Formula C22H24ClNO2S
Formula Weight 401.95
Synonym 5-Chloro-N-(6-phenylhexyl)-1-naphthalenesulfonamide
Accessories

Store at Room Temperature