Please login to your online account to display your discounted pricing

SC 79, Tocris™

Akt activator

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material SC 79
Assay Percent Range >99%
Molecular Formula C17H17ClN2O5
Formula Weight 364.78
Synonym 2-Amino-6-chloro-α-cyano-3-(ethoxycarbonyl)-4H-1-benzopyran-4-acetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

463550

 
r & d systems
RDS463550
50mg Each for $924.23

463510

 
r & d systems
RDS463510
10mg Each for $225.77
Description & Specifications

Specifications

Formula Weight 364.78
Quantity 50mg
Chemical Name or Material SC 79
Assay Percent Range >99%
Molecular Formula C17H17ClN2O5
CAS 305834-79-1
Synonym 2-Amino-6-chloro-α-cyano-3-(ethoxycarbonyl)-4H-1-benzopyran-4-acetic acid ethyl ester
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C