Please login to your online account to display your discounted pricing

SC 66, Tocris™

Allosteric Akt inhibitor

$121.23 - $509.13

Chemical Identifiers

Chemical Name or Material SC 66
Assay Percent Range >98%
Molecular Formula C18H16N2O
Formula Weight 276.33
Synonym (2E, 6E)-2, 6-Bis(4-pyridinylmethylene)cyclohexanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

439810

 
r & d systems
RDS439810
10mg Each for $121.23

439850

 
r & d systems
RDS439850
50mg Each for $509.13
Description & Specifications

Specifications

Formula Weight 276.33
Quantity 10mg
Recommended Storage Store at -20°C
CAS 871361-88-5
Assay Percent Range >98%
Chemical Name or Material SC 66
Molecular Formula C18H16N2O
Synonym (2E, 6E)-2, 6-Bis(4-pyridinylmethylene)cyclohexanone
Safety and Handling

Store at -20°C