Please login to your online account to display your discounted pricing

SC 58125, Tocris

Selective cyclooxygenase-2 (COX-2) inhibitor

$177.37 - $738.82

Chemical Identifiers

Chemical Name or Material SC 58125
Assay Percent Range >98%
Molecular Formula C17H12F4N2O2S
Formula Weight 384.35
Synonym 5-(4-Fluorophenyl)-1-[4-(methylsulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

289510

 
r & d systems
289510
10mg Each for $177.37

289550

 
r & d systems
289550
50mg Each for $738.82
Description & Specifications

Specifications

Formula Weight 384.35
Quantity 10mg
Chemical Name or Material SC 58125
Assay Percent Range >98%
Molecular Formula C17H12F4N2O2S
CAS 162054-19-5
Synonym 5-(4-Fluorophenyl)-1-[4-(methylsulfonyl)phenyl]-3-(trifluoromethyl)-1H-pyrazole
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C