Please login to your online account to display your discounted pricing

SC 57461A, Tocris

Selective inhibitor of LTA4 hydrolase

$188.60 - $794.96

Chemical Identifiers

Chemical Name or Material SC 57461A
Assay Percent Range >98%
Molecular Formula C20H25NO3·HCl
Formula Weight 363.88
Synonym N-methyl-N-[3-[4-(phenylmethyl)phenoxy]propyl]-β-alanine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

310710

 
r & d systems
RDS310710
10mg Each for $188.60

310750

 
r & d systems
RDS310750
50mg Each for $794.96
Description & Specifications

Specifications

Formula Weight 363.88
Quantity 10mg
Chemical Name or Material SC 57461A
CAS 423169-68-0
Synonym N-methyl-N-[3-[4-(phenylmethyl)phenoxy]propyl]-β-alanine hydrochloride
Assay Percent Range >98%
Molecular Formula C20H25NO3·HCl
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C