Please login to your online account to display your discounted pricing

Alfa Aesar™ SC-51322, 98%


Quantity 5mg

Manufacturer: alfa aesar™  J67002LB0

 View more versions of this product

Catalog No. AAJ67002LB0

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 457.94
MDL Number MFCD00916246
CAS 146032-79-3
Chemical Name or Material SC-51322
Assay 98%
RTECS Number HQ4002000
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Solid
UN Number UN2811
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Color White to off-white
Storage Note 1 -30° to -10°C
Quantity 5mg
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.

Specifications

Chemical Name or Material SC-51322
CAS 146032-79-3
Formula Weight 457.94
MDL Number MFCD00916246
UN Number UN2811
Accessories

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.

-30° to -10°C

SDS & Safety Documents