Please login to your online account to display your discounted pricing

SC-10, Tocris

Protein kinase C activator

$181.19 - $181.19

Chemical Identifiers

Chemical Name or Material SC-10
Assay Percent Range >98%
Molecular Formula C17H22ClNO2S
Formula Weight 339.88
Synonym 5-Chloro-N-heptylnaphthalene-1-sulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

043010

 
r & d systems
RDS043010
10mg Each for $181.19
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 339.88
Chemical Name or Material SC-10
Assay Percent Range >98%
CAS 102649-79-6
Synonym 5-Chloro-N-heptylnaphthalene-1-sulfonamide
Molecular Formula C17H22ClNO2S
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature