Please login to your online account to display your discounted pricing

SBE 13 hydrochloride, Tocris™

Potent and selective PLK1 inhibitor

$126.32 - $534.65

Chemical Identifiers

Chemical Name or Material SBE 13 hydrochloride
Assay Percent Range >99%
Molecular Formula C24H27ClN2O4·HCl
Formula Weight 479.4
Synonym N-[[4-[(6-Chloro-3-pyridinyl)methoxy]-3-methoxyphenyl]methyl]-3, 4-dimethoxybenzeneethanamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

429210

 
r & d systems
RDS429210
10mg Each for $126.32

429250

 
r & d systems
RDS429250
50mg Each for $534.65
Description & Specifications

Specifications

Formula Weight 479.4
Quantity 10mg
Chemical Name or Material SBE 13 hydrochloride
Assay Percent Range >99%
Molecular Formula C24H27ClN2O4·HCl
CAS 1052532-15-6
Synonym N-[[4-[(6-Chloro-3-pyridinyl)methoxy]-3-methoxyphenyl]methyl]-3, 4-dimethoxybenzeneethanamine hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature