Please login to your online account to display your discounted pricing

SB 611812, Tocris™

Urotensin-II (UT) antagonist; attenuates cardiac dysfunction

$244.81 - $997.23

Chemical Identifiers

Chemical Name or Material SB 611812
Assay Percent Range >99%
Molecular Formula C17H16Cl3F3N2O3S
Formula Weight 491.74
Synonym 2, 6-Dichloro-N-[4-chloro-3-[2-(dimethylamino)ethoxy]phenyl]-4-(trifluoromethyl)benzenesulfonamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

467050

 
r & d systems
RDS467050
50mg Each for $997.23

467010

 
r & d systems
RDS467010
10mg Each for $244.81
Description & Specifications

Specifications

Formula Weight 491.74
Quantity 50mg
CAS 345892-71-9
Chemical Name or Material SB 611812
Assay Percent Range >99%
Molecular Formula C17H16Cl3F3N2O3S
Synonym 2, 6-Dichloro-N-[4-chloro-3-[2-(dimethylamino)ethoxy]phenyl]-4-(trifluoromethyl)benzenesulfonamide
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C