Please login to your online account to display your discounted pricing

SB 590885, Tocris

Potent B-Raf inhibitor

$297.57 - $1,231.36

Chemical Identifiers

Chemical Name or Material SB 590885
Assay Percent Range >98%
Molecular Formula C27H27N5O2
Formula Weight 453.54
Synonym 5-[2-[4-[2-(Dimethylamino)ethoxy]phenyl]-5-(4-pyridinyl)-1H-imidazol-4-yl]-2, 3-dihydro-1H-inden-1-one oxime
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

265010

 
r & d systems
RDS265010
10mg Each for $297.57

265050

 
r & d systems
RDS265050
50mg Each for $1,231.36
Description & Specifications

Specifications

Formula Weight 453.54
Quantity 10mg
Chemical Name or Material SB 590885
Synonym 5-[2-[4-[2-(Dimethylamino)ethoxy]phenyl]-5-(4-pyridinyl)-1H-imidazol-4-yl]-2, 3-dihydro-1H-inden-1-one oxime
Molecular Formula C27H27N5O2
Assay Percent Range >98%
CAS 405554-55-4
Recommended Storage Store at -20°C
Safety and Handling

Store at -20°C