Please login to your online account to display your discounted pricing

SB 452533, Tocris

Potent TRPV1 receptor antagonist

$199.70 - $834.54

Chemical Identifiers

Chemical Name or Material SB 452533
Assay Percent Range >98%
Molecular Formula C18H22BrN3O
Formula Weight 376.29
Synonym N-(2-Bromophenyl)-N'-[2-[ethyl(3-methylphenyl)amino]ethyl]-urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

326510

 
r & d systems
RDS326510
10mg Each for $199.70

326550

 
r & d systems
RDS326550
50mg Each for $834.54
Description & Specifications

Specifications

Formula Weight 376.29
Quantity 10mg
Chemical Name or Material SB 452533
Assay Percent Range >98%
Molecular Formula C18H22BrN3O
CAS 459429-39-1
Synonym N-(2-Bromophenyl)-N'-[2-[ethyl(3-methylphenyl)amino]ethyl]-urea
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature