Please login to your online account to display your discounted pricing

SB 415286, Tocris

Potent, selective GSK-3 inhibitor

$238.81 - $983.93

Chemical Identifiers

Chemical Name or Material SB 415286
Assay Percent Range >99%
Molecular Formula C16H10ClN3O5
Formula Weight 359.73
Synonym 3-[(3-Chloro-4-hydroxyphenyl)amino]-4-(2-nitrophenyl)-1H-pyrrole-2, 5-dione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

161710

 
r & d systems
161710
10mg Each for $238.81

161750

 
r & d systems
161750
50mg Each for $983.93
Description & Specifications

Specifications

Formula Weight 359.73
Quantity 10mg
Chemical Name or Material SB 415286
Assay Percent Range >99%
Molecular Formula C16H10ClN3O5
CAS 264218-23-7
Synonym 3-[(3-Chloro-4-hydroxyphenyl)amino]-4-(2-nitrophenyl)-1H-pyrrole-2, 5-dione
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature