Please login to your online account to display your discounted pricing

SB 399885 hydrochloride, Tocris

Potent and selective 5-HT6 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material SB 399885 hydrochloride
Assay Percent Range >99%
Molecular Formula C18H21Cl2N3O4S·HCl
Formula Weight 482.81
Synonym N-(3, 5-Dichloro-2-methoxyphenyl)-4-methoxy-3-(1-piperazinyl)benzenesulfonamide hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

318950

 
r & d systems
RDS318950
50mg Each for $1,050.42

318910

 
r & d systems
RDS318910
10mg Each for $252.39
Description & Specifications

Specifications

Formula Weight 482.81
Quantity 50mg
Recommended Storage Desiccate at Room Temperature
Molecular Formula C18H21Cl2N3O4S·HCl
Assay Percent Range >99%
Chemical Name or Material SB 399885 hydrochloride
Synonym N-(3, 5-Dichloro-2-methoxyphenyl)-4-methoxy-3-(1-piperazinyl)benzenesulfonamide hydrochloride
CAS 402713-81-9
Safety and Handling

Desiccate at Room Temperature