Please login to your online account to display your discounted pricing

SB 366791, Tocris

Potent, selective, competitive TRPV1 (VR1) antagonist

$190.13 - $816.90

Chemical Identifiers

Chemical Name or Material SB 366791
Assay Percent Range >99%
Molecular Formula C16H14ClNO2
Formula Weight 287.75
Synonym 4'-Chloro-3-methoxycinnamanilide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

161510

 
r & d systems
RDS161510
10mg Each for $190.13

161550

 
r & d systems
RDS161550
50mg Each for $816.90
Description & Specifications

Specifications

Formula Weight 287.75
Quantity 10mg
Molecular Formula C16H14ClNO2
Assay Percent Range >99%
Synonym 4'-Chloro-3-methoxycinnamanilide
CAS 472981-92-3
Chemical Name or Material SB 366791
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature