Please login to your online account to display your discounted pricing

Alfa Aesar™ SB-366791, 99%

Chemical Identifiers

Chemical Name or Material SB-366791
CAS 472981-92-3
Formula Weight 287.75
MDL Number MFCD01033818
Synonym 4-Chloro-N-(3-methoxyphenyl)cinnamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67312LB0

 
alfa aesar™
J67312LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 287.75
Melting Point 165° to 167°C
CAS 472981-92-3
MDL Number MFCD01033818
Chemical Name or Material SB-366791
Synonym 4-Chloro-N-(3-methoxyphenyl)cinnamide
Assay 99%
Color Pale brown
Physical Form Solid
Quantity 5mg