Please login to your online account to display your discounted pricing

SB 334867, Tocris

Selective non-peptide OX1 antagonist

$252.39 - $1,050.42

Chemical Identifiers

Chemical Name or Material SB 334867
Assay Percent Range >99%
Molecular Formula C17H13N5O2
Formula Weight 319.32
Synonym N-(2-Methyl-6-benzoxazolyl)-N'-1, 5-naphthyridin-4-yl urea
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

19-601-0

 
r & d systems
196010
10mg Each for $252.39

19-605-0

 
r & d systems
196050
50mg Each for $1,050.42
Description & Specifications

Specifications

Formula Weight 319.32
Quantity 10mg
Chemical Name or Material SB 334867
Synonym N-(2-Methyl-6-benzoxazolyl)-N'-1, 5-naphthyridin-4-yl urea
Molecular Formula C17H13N5O2
Assay Percent Range >99%
Recommended Storage Desiccate at Room Temperature
CAS 792173-99-0
Safety and Handling

Desiccate at Room Temperature