Please login to your online account to display your discounted pricing

Tocris™ SB 328437

Potent and selective CCR3 antagonist

$218.74 - $915.10

Specifications

Product Type SB 328437
Inhibitors SB 328437
Molecular Weight 378.38
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

365010R

 
tocris™
RDS365010
10mg Each for $218.74

365050R

 
tocris™
RDS365050
50mg Each for $915.10
Description & Specifications

Specifications

Product Type SB 328437
Inhibitors SB 328437
Molecular Weight 378.38
Purity >99

Potent and selective CCR3 antagonist (IC50 = 4nM). Displays > 2500-fold selectivity over C5aR, LTD4, CCR7, CXCR1 and CXCR2 receptors. Inhibits eotaxin-, eotaxin-2- and MCP-4-induced Ca2+ mobilization (IC50 values are 38, 35 and 20nM respectively) and inhibits eotaxin-, eotaxin-2- and MCP-4-induced eosinophil chemotaxis with similar potencies.

  • Chemical Name: N-(1-Naphthalenylcarbonyl)-4-nitro-L-phenylalanine methyl ester
  • Formula: C21H18N2O5
  • CAS Number: 247580-43-4
  • Solubility: Soluble to 100mM in DMSO
  • Purity: >99