Please login to your online account to display your discounted pricing

Tocris™ SB 297006

Potent and selective CCR3 antagonist

$221.01 - $904.70

Specifications

Product Type SB 297006
Inhibitors SB 297006
Molecular Weight 342.35
Purity >99
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

421310

 
tocris™
RDS421310
10mg Each for $221.01

421350

 
tocris™
RDS421350
50mg Each for $904.70
Description & Specifications

Specifications

Product Type SB 297006
Inhibitors SB 297006
Molecular Weight 342.35
Purity >99

Potent and selective CCR3 antagonist (IC50 = 39nM). Displays 250-fold selectivity for CCR3 over other chemokine receptors, including CXCR1, CXCR2, CCR1 and CCR7 (IC50 >27μM). Inhibits calcium mobilization induced by MCP-4, eotaxin-2 and eotaxin in RBL-2H3 cells transfected with CCR3 (IC50 values are 80, 90 and 210nM respectively).

  • Chemical Name: N-Benzoyl-4-nitroaniline ethyl ester
  • Formula: C18H18N2O5
  • CAS Number: 58816-69-6
  • Solubility: Soluble to 100mM in DMSO and to 20mM in ethanol
  • Purity: >99