Please login to your online account to display your discounted pricing

SB 269970 hydrochloride, Tocris

Potent selective 5-HT7 antagonist; Brain penetrant

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material SB 269970 hydrochloride
Assay Percent Range >98%
Molecular Formula C18H28N2O3S·HCl
Formula Weight 388.95
Synonym (2R)-1-[(3-Hydroxyphenyl)sulfonyl]-2-[2-(4-methyl-1-piperidinyl)ethyl]pyrrolidine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

161250

 
r & d systems
RDS161250
50mg Each for $1,073.10

161210

 
r & d systems
RDS161210
10mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 388.95
Quantity 50mg
Molecular Formula C18H28N2O3S·HCl
CAS 261901-57-9
Chemical Name or Material SB 269970 hydrochloride
Assay Percent Range >98%
Synonym (2R)-1-[(3-Hydroxyphenyl)sulfonyl]-2-[2-(4-methyl-1-piperidinyl)ethyl]pyrrolidine hydrochloride
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C