missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SB 258585 hydrochloride, Tocris

Potent, selective 5-HT6 antagonist

$218.74 - $915.10

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C18H22IN3O3S·HCl
Formula Weight 523.82
Synonym 4-Iodo-N-[4-methoxy-3-(4-methyl-1-piperazinyl)phenyl]benzenesulfonamide hydrochloride
Chemical Name or Material SB 258585 hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
196110 R & D Systems
RDS196110
10mg Each for $218.74
196150 R & D Systems
RDS196150
50mg Each for $915.10
Specifications Description & Specifications

Chemical Identifiers

>99%
523.82
SB 258585 hydrochloride
C18H22IN3O3S·HCl
4-Iodo-N-[4-methoxy-3-(4-methyl-1-piperazinyl)phenyl]benzenesulfonamide hydrochloride

Specifications

1216468-02-8
C18H22IN3O3S·HCl
4-Iodo-N-[4-methoxy-3-(4-methyl-1-piperazinyl)phenyl]benzenesulfonamide hydrochloride
SB 258585 hydrochloride
>99%
523.82
10mg

Safety and Handling

Recommended Storage : Desiccate at +4°C