Please login to your online account to display your discounted pricing

SB 242084, Tocris

Selective 5-HT2C antagonist; brain penetrant

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material SB 242084
Assay Percent Range >99%
Molecular Formula C21H19ClN4O2·2HCl
Formula Weight 467.78
Synonym 6-Chloro-2, 3-dihydro-5-methyl-N-[6-[(2-methyl-3-pyridinyl)oxy]-3-pyridinyl]-1H-indole-1-carboxyamide dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

290150

 
r & d systems
290150
50mg Each for $1,073.10

290110

 
r & d systems
290110
10mg Each for $257.85
Description & Specifications

Specifications

Formula Weight 467.78
Quantity 50mg
Chemical Name or Material SB 242084
Assay Percent Range >99%
Molecular Formula C21H19ClN4O2·2HCl
CAS 1215566-78-1
Synonym 6-Chloro-2, 3-dihydro-5-methyl-N-[6-[(2-methyl-3-pyridinyl)oxy]-3-pyridinyl]-1H-indole-1-carboxyamide dihydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C